Learn More
Medchemexpress LLC Dihydroergotamine mesylate | 6190-39-2 | 99.9% | 5 MG
SDP
Catalog No. 5000408415
Encompass_Preferred
Dihydroergotamine mesylate is a blood-brain barrier (BBB)-permeable ergot alkaloid used in migraine research. It targets 5-HT Receptors within GPCR/G Protein and neuronal signaling pathways.
  • Acts as a 5-HT receptor agonist.
  • Reduces cell survival in A549 and NCI-H226 cells.
  • Inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells.
  • Induces mitochondrial morphologic alterations and dysfunction.
  • Increases reactive oxygen species (ROS) generation.
  • Triggers apoptosis and disrupts ATP production.
  • Inhibits STAT3 activation and enhances Mcl-1 protein stability.
  • Inhibits tumor growth in HepG2 xenografted mice.
  • Reduces Ki67 positive cells, Mcl-1, p-Mcl-1, and p-ERK expression.
Catalog No. 50-004-08415 Supplier Medchemexpress LLC Supplier No. HYB0670A5MG
Dihydroergotamine mesylate is a blood-brain barrier (BBB)-permeable ergot alkaloid used in migraine research. It targets 5-HT Receptors within GPCR/G Protein and neuronal signaling pathways.
  • Acts as a 5-HT receptor agonist.
  • Reduces cell survival in A549 and NCI-H226 cells.
  • Inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells.
  • Induces mitochondrial morphologic alterations and dysfunction.
  • Increases reactive oxygen species (ROS) generation.
  • Triggers apoptosis and disrupts ATP production.
  • Inhibits STAT3 activation and enhances Mcl-1 protein stability.
  • Inhibits tumor growth in HepG2 xenografted mice.
  • Reduces Ki67 positive cells, Mcl-1, p-Mcl-1, and p-ERK expression.