Learn More
Medchemexpress LLC Dihydroergotamine mesylate | 6190-39-2 | 99.94% | 50 MG
SDP
Catalog No. 5000409045
Encompass_Preferred
Dihydroergotamine mesylate is a BBB-permeable ergot alkaloid for the research of migraines. It reduces cell survival in A549 and NCI-H226 cells and inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells.
  • Reduces cell survival in A549 and NCI-H226 cells
  • Inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells
  • Induces mitochondrial morphologic alterations and dysfunction
  • Increases ROS generation and induces apoptosis
  • Disturbs ATP production in A549 cells
  • Inhibits STAT3 activation and enhances protein stability of Mcl-1
Catalog No. 50-004-09045 Supplier Medchemexpress LLC Supplier No. HYB0670A50MG
Dihydroergotamine mesylate is a BBB-permeable ergot alkaloid for the research of migraines. It reduces cell survival in A549 and NCI-H226 cells and inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells.
  • Reduces cell survival in A549 and NCI-H226 cells
  • Inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells
  • Induces mitochondrial morphologic alterations and dysfunction
  • Increases ROS generation and induces apoptosis
  • Disturbs ATP production in A549 cells
  • Inhibits STAT3 activation and enhances protein stability of Mcl-1