Learn More
Cayman Chemical DIhydroergotamInmesylate 100mg
SDP
Catalog No. 502994804
Encompass_Preferred
An agonist of 5-HT1B and 5-HT1D receptors (Kis = 0.3 and 2.5 nM, respectively, for human receptors); binds to dopamine D2 and D3 as well as 5-HT2A receptors (K0.5s = 5, 16, and 38 nM, respectively) but has lower affinity for D1, 5-HT2C, and 5-HT3 receptors (K0.5s = 2,779, 298, and >10,000 nM, respectively); binds to α-adrenergic receptors; increases the mechanical pain threshold in a rat model of neuropathic pain when used at doses of 50 and 100 mg/kg
Catalog No. 50-299-4804 Supplier Cayman Chemical Supplier No. 23847100 MG
An agonist of 5-HT1B and 5-HT1D receptors (Kis = 0.3 and 2.5 nM, respectively, for human receptors); binds to dopamine D2 and D3 as well as 5-HT2A receptors (K0.5s = 5, 16, and 38 nM, respectively) but has lower affinity for D1, 5-HT2C, and 5-HT3 receptors (K0.5s = 2,779, 298, and >10,000 nM, respectively); binds to α-adrenergic receptors; increases the mechanical pain threshold in a rat model of neuropathic pain when used at doses of 50 and 100 mg/kg