Learn More
Selleck Chemical LLC Dihydroethidium
SDP
Catalog No. 5000002590
Encompass_Preferred
Dihydroethidium (DHE HE Hydroethidine PD-MY 003) is a cell-permeable blue fluorogenic probe used for detecting intracellular superoxide radical anion
Catalog No. 50-000-02590 Supplier Selleck Chemical LLC Supplier No. S67925MG
Dihydroethidium (DHE HE Hydroethidine PD-MY 003) is a cell-permeable blue fluorogenic probe used for detecting intracellular superoxide radical anion