Learn More
Selleck Chemical LLC Dihydroethidium
SDP
Supplier:  Selleck Chemical LLC S679225MG
Encompass_Preferred
Dihydroethidium (DHE HE Hydroethidine PD-MY 003) is a cell-permeable blue fluorogenic probe used for detecting intracellular superoxide radical anion
Catalog No. 50-000-02591