Learn More
Cayman Chemical Dihydroethidium, Purity: ≥98%, CAS: 104821-25-2, 5 mg
SDP
Catalog No. 502955964
Encompass_Preferred
A fluorescent dye to monitor superoxide production; ex/em = 490/590 nm; has been used to detect ROS and intracellular superoxide in vitro
This item is not returnable. View return policy
Catalog No. 50-295-5964 Supplier Cayman Chemical Supplier No. 120135 MG
A fluorescent dye to monitor superoxide production; ex/em = 490/590 nm; has been used to detect ROS and intracellular superoxide in vitro