Learn More
Apexbio Technology LLC Dihydroethidium(Synonyms: Hydroethidine, DHE, HE, Dihydroethidine, Ethidium hydride), 5mg, CAS: 104821-25-2.
SDP
Catalog No. 5000060446
Encompass_Preferred
Dihydroethidium (CAS 104821-25-2) also known as hydroethidine is a cell-permeable fluorescent probe widely utilized for detecting superoxide anions (O ) in live cells After entering cells dihydroethidium is oxidized by intracellular superoxide anions to ethidium which subsequently intercalates into DNA emitting detectable red fluorescence (excitation/emission 518/605 nm) Unoxidized dihydroethidium exhibits blue fluorescence (355/420 nm) The intensity of the red fluorescence correlates directly with intracellular O concentration Thus dihydroethidium serves as a valuable tool in evaluating oxidative stress and studying associated physiological/pathological processes including apoptosis cell proliferation cardiovascular pathologies diabetes and cancer
Catalog No. 50-000-60446 Supplier Apexbio Technology LLC Supplier No. C38075
Dihydroethidium (CAS 104821-25-2) also known as hydroethidine is a cell-permeable fluorescent probe widely utilized for detecting superoxide anions (O ) in live cells After entering cells dihydroethidium is oxidized by intracellular superoxide anions to ethidium which subsequently intercalates into DNA emitting detectable red fluorescence (excitation/emission 518/605 nm) Unoxidized dihydroethidium exhibits blue fluorescence (355/420 nm) The intensity of the red fluorescence correlates directly with intracellular O concentration Thus dihydroethidium serves as a valuable tool in evaluating oxidative stress and studying associated physiological/pathological processes including apoptosis cell proliferation cardiovascular pathologies diabetes and cancer