Learn More
AdipoGen Dihydroethidium
SDP
Catalog No. 502561244
Encompass_Preferred
Chemical. CAS 104821-25-2. Formula C21H21N3. MW 315.41. Synthetic. Redox indicator. Blue fluorescence until oxidized to ethidium. Can be used for detecting superoxide radical in cells, tissues and organisms.
Catalog No. 50-256-1244 Supplier AdipoGen Supplier No. CDXD0094M100
Chemical. CAS 104821-25-2. Formula C21H21N3. MW 315.41. Synthetic. Redox indicator. Blue fluorescence until oxidized to ethidium. Can be used for detecting superoxide radical in cells, tissues and organisms.