Learn More
Abcam Dihydroethidium (DHE), Blue fluorescent dye, 100MG
Catalog No. 502705264
Encompass_Preferred
MW 315.4 Da. Cell-permeable blue fluorescent dye. Redox indicator. Loses fluorescence on oxidation to ethidium (ab141391). Detects superoxide radical in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5264 Supplier Abcam Supplier No. AB145360100MG
MW 315.4 Da. Cell-permeable blue fluorescent dye. Redox indicator. Loses fluorescence on oxidation to ethidium (ab141391). Detects superoxide radical in vivo. .
The product is subject to the following: Abcam Restricted Use Statement