Learn More
Medchemexpress LLC HY-N7080 100mg Medchemexpress, Dihydroferulic acid CAS:1135-23-5 Purity:>98%
SDP
Catalog No. 501970502
Encompass_Preferred
Medchemexpress, HY-N7080 100mg Dihydroferulic acid CAS:1135-23-5 Dihydroferulic acid (Hydroferulic acid) is one of the main metabolites of curcumin and antioxidant/radical-scavenging properties with an IC50 value of 19.5 μM. Dihydroferulic acid is a metabolite of human gut microflora as well as a precursor of vanillic acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0502 Supplier Medchemexpress LLC Supplier No. HYN7080100MG
Medchemexpress, HY-N7080 100mg Dihydroferulic acid CAS:1135-23-5 Dihydroferulic acid (Hydroferulic acid) is one of the main metabolites of curcumin and antioxidant/radical-scavenging properties with an IC50 value of 19.5 μM. Dihydroferulic acid is a metabolite of human gut microflora as well as a precursor of vanillic acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.