Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrofisetin, MP Biomedicals

Catalog No. ICN210015 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210015 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210015 Supplier MP Biomedicals, Inc Supplier No. 0521001510

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20725-03-5
Molecular Formula C15H12O6
Molecular Weight (g/mol) 288.255
InChI Key FNUPUYFWZXZMIE-LSDHHAIUSA-N
Synonym fustin, 2,3-dihydrofisetin, fustin, +, unii-c308qr79ue, 3,7,3',4'-tetrahydroxyflavanone, tetrahydroxyflavanone, +-fustin, fustin 2r,3r-form mi, flavanone, 3,3',4',7-tetrahydroxy-, +, 3,7,3',4'-tetrahydroxy-flavanon
PubChem CID 5317435
IUPAC Name (2R,3R)-2-(3,4-dihydroxyphenyl)-3,7-dihydroxy-2,3-dihydrochromen-4-one
SMILES C1=CC(=C(C=C1C2C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O

Specifications

Quantity 100 mg
Formula Weight 288.26
Chemical Name or Material Dihydrofisetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.