Learn More
Medchemexpress LLC Dihydrofluorescein diacetate | 35340-49-9 | 98.5% | 418.40 g/mol | C24H18O7 | 1 ML
SDP
Catalog No. 5000245568
Encompass_Preferred
Dihydrofluorescein diacetate is a cell-permeable fluorimetric probe used to detect reactive oxygen species and assess oxidative stress in cell-free systems and live cells. Intracellular esterases remove the diacetate groups to generate the reduced fluorophore, which is oxidized by oxidants to produce a measurable fluorescent signal.
  • Cell-permeable fluorimetric probe for oxidative stress measurements.
  • Hydrolyzed by intracellular esterases to yield a fluorescent product.
  • Applicable to both cell-free and cell-based assays.
  • Supplied as a 10 mM stock solution in DMSO for convenient dosing.
  • Excitation ~518 nm; emission ~616 nm for fluorescence detection.
  • High purity (~98.5%) for reproducible experimental results.
  • Store protected from light; solution stability: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-45568 Supplier Medchemexpress LLC Supplier No. HY10189310MM 1ML
Dihydrofluorescein diacetate is a cell-permeable fluorimetric probe used to detect reactive oxygen species and assess oxidative stress in cell-free systems and live cells. Intracellular esterases remove the diacetate groups to generate the reduced fluorophore, which is oxidized by oxidants to produce a measurable fluorescent signal.
  • Cell-permeable fluorimetric probe for oxidative stress measurements.
  • Hydrolyzed by intracellular esterases to yield a fluorescent product.
  • Applicable to both cell-free and cell-based assays.
  • Supplied as a 10 mM stock solution in DMSO for convenient dosing.
  • Excitation ~518 nm; emission ~616 nm for fluorescence detection.
  • High purity (~98.5%) for reproducible experimental results.
  • Store protected from light; solution stability: -80°C up to 6 months, -20°C up to 1 month.