Learn More
Aat Bioquest Dihydrofluorescein diacetate [Fluorescin diacetate] *CAS 35340-49-9*
SDP
Catalog No. 501953550
Encompass_Preferred
Dihydrofluorescein diacetate (also called fluorescin diacetate) is hydrolyzed by cellular esterases to dihydrofluorescein (also called fluorescin) and is then oxidized to fluorescein primarily by H2O2.
Catalog No. 50-195-3550 Supplier Aat Bioquest Supplier No. 15203
Dihydrofluorescein diacetate (also called fluorescin diacetate) is hydrolyzed by cellular esterases to dihydrofluorescein (also called fluorescin) and is then oxidized to fluorescein primarily by H2O2.