Learn More
AdipoGen Dihydrofluorescein diacetate
SDP
Catalog No. 502561227
Encompass_Preferred
Chemical. CAS 35340-49-9. Formula C24H18O7. MW 418.4. Synthetic. Dihydrofluorescein diacetate is a fluorescent probe for detecting intracellular oxidants. It is hydrolyzed by cellular esterases to dihydrofluorescein and is then oxidized to fluorescein primarily by H2O2. Dihydrofluorescein diacetate might be reactive toward a broad range of oxidizing reactions that may be increased during intracellular oxidant stress. Cell-loading studies indicate that dihydrofluorescein achieves higher intracellular concentrations than the other redox sensors such as Dihydrorhodamine 123 Ex/Em 490/514nm.
Catalog No. 50-256-1227 Supplier AdipoGen Supplier No. CDXD0122G010
Chemical. CAS 35340-49-9. Formula C24H18O7. MW 418.4. Synthetic. Dihydrofluorescein diacetate is a fluorescent probe for detecting intracellular oxidants. It is hydrolyzed by cellular esterases to dihydrofluorescein and is then oxidized to fluorescein primarily by H2O2. Dihydrofluorescein diacetate might be reactive toward a broad range of oxidizing reactions that may be increased during intracellular oxidant stress. Cell-loading studies indicate that dihydrofluorescein achieves higher intracellular concentrations than the other redox sensors such as Dihydrorhodamine 123 Ex/Em 490/514nm.