Learn More
Abcam Dihydrofluorescein diacetate, Fluorescent intracellular o x idant probe, 10G
Catalog No. 502705271
Encompass_Preferred
MW 418.4 Da, Purity >97%. Fluorescent intracellular oxidant probe. Redox sensor. Achieves higher intracellular concentrations resulting in brighter intracellular labelling but is less bright extracellularly than dihydrorhodamine 123 (ab145260). Oxidizes to fluorescein primarily under H₂O₂.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5271 Supplier Abcam Supplier No. AB14535110G
MW 418.4 Da, Purity >97%. Fluorescent intracellular oxidant probe. Redox sensor. Achieves higher intracellular concentrations resulting in brighter intracellular labelling but is less bright extracellularly than dihydrorhodamine 123 (ab145260). Oxidizes to fluorescein primarily under H₂O₂.
The product is subject to the following: Abcam Restricted Use Statement