Learn More
Sigma Aldrich Fine Chemicals Biosciences Dihydrofolate Reductase hu
Catalog No. 502905457
Encompass
Human DHfR is an 186 amino acid protein with an apparent molecular weight of 25 kDa. It is 30% homologous to the E. coli protein and up to 70% homologous to vertebrate proteins.
Catalog No. 50-290-5457 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D6566.25UN
Human DHfR is an 186 amino acid protein with an apparent molecular weight of 25 kDa. It is 30% homologous to the E. coli protein and up to 70% homologous to vertebrate proteins.