Learn More
Cayman Chemical DIhydrofolIc AcId 10mg
SDP
Catalog No. 502947358
Encompass_Preferred
An active metabolite of folic acid; formed via the NADH-dependent reduction of folic acid by DHFR; reduces IGF-1R promoter activity in a cell-based reporter assay at 10, 50, and 100 UG/ml
Catalog No. 50-294-7358 Supplier Cayman Chemical Supplier No. 3275010 MG
An active metabolite of folic acid; formed via the NADH-dependent reduction of folic acid by DHFR; reduces IGF-1R promoter activity in a cell-based reporter assay at 10, 50, and 100 UG/ml