Learn More
Medchemexpress LLC Dihydrofuran-3(2H)-one | 22929-52-8 | 100.0% | 86.09 | 250 G
SDP
Catalog No. 5000402334
Encompass_Preferred
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It is a building block for the synthesis of biologically active compounds.
  • Used to synthesize cyclic ketone inhibitors
  • Inhibits serine protease plasmin
  • Building block for biologically active compounds
Catalog No. 50-004-02334 Supplier Medchemexpress LLC Supplier No. 250GHY33900
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It is a building block for the synthesis of biologically active compounds.
  • Used to synthesize cyclic ketone inhibitors
  • Inhibits serine protease plasmin
  • Building block for biologically active compounds