Learn More
Medchemexpress LLC Dihydrofuran-3(2H)-one | 22929-52-8 | 99.97% | 86.09 | 25 G
SDP
Catalog No. 5000403003
Encompass_Preferred
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It is also a building block for the synthesis of biologically active compounds. This product is suitable for laboratory research and development.
  • Used to synthesize cyclic ketone inhibitors
  • Inhibits serine protease plasmin
  • Building block for biologically active compounds
  • Suitable for research and development
Catalog No. 50-004-03003 Supplier Medchemexpress LLC Supplier No. 25GHY33900
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It is also a building block for the synthesis of biologically active compounds. This product is suitable for laboratory research and development.
  • Used to synthesize cyclic ketone inhibitors
  • Inhibits serine protease plasmin
  • Building block for biologically active compounds
  • Suitable for research and development