Learn More
Medchemexpress LLC Dihydrofuran-3(2H)-one | 22929-52-8 | 99.99% | 86.09 | 10 G
SDP
Catalog No. 5000404467
Encompass_Preferred
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone primarily used as a building block for the synthesis of biologically active compounds. It is also utilized in the synthesis of cyclic ketone inhibitors that inhibit the serine protease plasmin.
  • Cyclic ketone structure
  • Building block for biologically active compounds
  • Inhibits serine protease plasmin
  • For research use only
Catalog No. 50-004-04467 Supplier Medchemexpress LLC Supplier No. HY3390010G
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone primarily used as a building block for the synthesis of biologically active compounds. It is also utilized in the synthesis of cyclic ketone inhibitors that inhibit the serine protease plasmin.
  • Cyclic ketone structure
  • Building block for biologically active compounds
  • Inhibits serine protease plasmin
  • For research use only