Learn More
Medchemexpress LLC Dihydrofuran-3(2H)-one | 22929-52-8 | 99.99% | 86.09 | 500 G
SDP
Catalog No. 5000404171
Encompass_Preferred
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It serves as a building block for the synthesis of biologically active compounds and is intended for research use only.
  • Cyclic ketone structure.
  • Can be used to synthesize cyclic ketone inhibitors.
  • Inhibits serine protease plasmin.
  • Building block for synthesizing biologically active compounds.
  • Suitable for research use.
Catalog No. 50-004-04171 Supplier Medchemexpress LLC Supplier No. HY33900500G
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It serves as a building block for the synthesis of biologically active compounds and is intended for research use only.
  • Cyclic ketone structure.
  • Can be used to synthesize cyclic ketone inhibitors.
  • Inhibits serine protease plasmin.
  • Building block for synthesizing biologically active compounds.
  • Suitable for research use.