Learn More
Medchemexpress LLC Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) | 22929-52-8 | 100.0% | 86.09 | 50 G
SDP
Catalog No. 5000404172
Encompass_Preferred
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone.
  • Used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin.
  • Building block for the synthesis of biologically active compounds.
Catalog No. 50-004-04172 Supplier Medchemexpress LLC Supplier No. HY3390050G
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone.
  • Used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin.
  • Building block for the synthesis of biologically active compounds.