Learn More
Medchemexpress LLC Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) | 22929-52-8 | 99.99% | 86.09 | 100 G
SDP
Catalog No. 5000404466
Encompass_Preferred
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It is also a building block for the synthesis of biologically active compounds.
  • Used to synthesize cyclic ketone inhibitors
  • Inhibits the serine protease plasmin
  • Building block for biologically active compounds
  • For research use only
Catalog No. 50-004-04466 Supplier Medchemexpress LLC Supplier No. HY33900100G
Dihydrofuran-3(2H)-one (3-Oxotetrahydrofuran) is a cyclic ketone that can be used to synthesize cyclic ketone inhibitors that inhibit the serine protease plasmin. It is also a building block for the synthesis of biologically active compounds.
  • Used to synthesize cyclic ketone inhibitors
  • Inhibits the serine protease plasmin
  • Building block for biologically active compounds
  • For research use only