Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dihydrofuran-3(2h)-one, 95%, Thermo Scientific™

Catalog No. MO08444DA
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO08444DA 1 g Amber Glass Bottle
MO08444DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO08444DA Supplier Thermo Scientific Chemicals Supplier No. MO08444DA
Add to Cart

Chemical Identifiers

CAS 22929-52-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD07778393
InChI Key JLPJFSCQKHRSQR-UHFFFAOYSA-N
Synonym 3-oxotetrahydrofuran, dihydrofuran-3 2h-one, dihydro-3 2h-furanone, dihydrofuran-3-one, dihydro-furan-3-one, tetrahydrofuran-3-one, 3 2h-furanone, dihydro, 3-oxo-1,4-epoxybutane, tetrahydrofuran-3-yloxy
PubChem CID 529392
IUPAC Name oxolan-3-one
SMILES C1COCC1=O

Specifications

CAS Min % 95.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 95%
Formula Weight 86.09
Percent Purity 95%
Chemical Name or Material Dihydrofuran-3(2H)-one
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.