Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrogen Di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate 97.0+%, TCI America™
SDP

Catalog No. D3706100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3706100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3706100MG Supplier TCI America Supplier No. D3706100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 386706-31-6
Molecular Formula C32H76Cl2O4P4Pd2-2
Molecular Weight (g/mol) 932.591
MDL Number MFCD05662696
InChI Key WJNABTRGRXAOEB-UHFFFAOYSA-N
PubChem CID 131674722
IUPAC Name ditert-butylphosphinite;ditert-butylphosphinous acid;palladium;dihydrochloride
SMILES CC(C)(C)P(C(C)(C)C)O.CC(C)(C)P(C(C)(C)C)O.CC(C)(C)P(C(C)(C)C)[O-].CC(C)(C)P(C(C)(C)C)[O-].Cl.Cl.[Pd].[Pd]

Specifications

Melting Point 280°C
Quantity 100 mg
Formula Weight 930.58
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dihydrogen Di-mu-chlorotetrakis(di-tert-butylphosphinito)dipalladate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.