Learn More
Cayman Chemical Dihydrojasmonic Acid, Purity: ≥95% (mixture of isomers), CAS: 3572-64-3, 250 mg
SDP
Catalog No. 502913644
Encompass_Preferred
A plant growth regulator; inhibits lettuce seed germination and reduces the radicle length of germinated seedlings at 10 mM
Catalog No. 50-291-3644 Supplier Cayman Chemical Supplier No. 35362250 MG
A plant growth regulator; inhibits lettuce seed germination and reduces the radicle length of germinated seedlings at 10 mM