Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrojasmonic acid | 3572-64-3 | 99.3% | 50 MG
SDP

Catalog No. 5000338651
Encompass_Preferred

Dihydrojasmonic acid is a plant growth regulator. It is intended for research use only and not sold to patients.

  • Plant growth regulator
  • For research use only
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Density: 1.038±0.06 g/cm3

Catalog No. 50-003-38651 Supplier Medchemexpress LLC Supplier No. HY13111650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Dihydrojasmonic acid is a plant growth regulator. It is intended for research use only and not sold to patients.

  • Plant growth regulator
  • For research use only
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Density: 1.038±0.06 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.