Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrojasmonic Acid 98.0+%, TCI America™
SDP

Catalog No. D32255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32255G 5 g
D32251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32255G Supplier TCI America Supplier No. D32255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3572-64-3
Molecular Formula C12H20O3
Molecular Weight (g/mol) 212.29
MDL Number MFCD18781916
InChI Key PQEYTAGBXNEUQL-UHFFFAOYNA-N
Synonym 2-Amyl-3-(carboxymethyl)cyclopentanone, 2-Amyl-3-oxocyclopentaneacetic Acid, 3-(Carboxymethyl)-2-pentylcyclopentanone, 3-Oxo-2-pentylcyclopentaneacetic Acid
PubChem CID 107126
IUPAC Name 2-(3-oxo-2-pentylcyclopentyl)acetic acid
SMILES CCCCCC1C(CC(O)=O)CCC1=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 212.29
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Dihydrojasmonic Acid
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.