Learn More
AdipoGen Dihydrokaempferol
SDP
Catalog No. 502561076
Encompass_Preferred
Chemical. CAS 104486-98-8. Formula C15H12O6. MW 288.25. Synthetic. Flavanonol derived from plant source. Precursor of kaempferol. Shown to stimulate glucose uptake and improve insulin resistance by inducing adipogenesis through increased PPAR2 expression. Anti-inflammatory compound.
Catalog No. 50-256-1076 Supplier AdipoGen Supplier No. CDXD0442M010
Chemical. CAS 104486-98-8. Formula C15H12O6. MW 288.25. Synthetic. Flavanonol derived from plant source. Precursor of kaempferol. Shown to stimulate glucose uptake and improve insulin resistance by inducing adipogenesis through increased PPAR2 expression. Anti-inflammatory compound.