Learn More
Cayman Chemical Dihydrokainic Acid, Purity: ≥95%, CAS: 52497-36-6, 10 mg
SDP
Catalog No. 502939078
Encompass_Preferred
An inhibitor of EAAT2 (Ki = 23 µM); selective for EAAT2 over EAAT1 and EAAT3 (Ki = >3 mM for both); reduces depressive-like behavior in rats in the forced swim test at 5 nmol infused into the infralimbic cortex; induces anhedonia-like behavior in rats in a sucrose intake test at 6.25 nmol infused into the PFC; impairs memory acquisition, consolidation, and retrieval in mice in the novel object recognition test
Catalog No. 50-293-9078 Supplier Cayman Chemical Supplier No. 2847810 MG
An inhibitor of EAAT2 (Ki = 23 µM); selective for EAAT2 over EAAT1 and EAAT3 (Ki = >3 mM for both); reduces depressive-like behavior in rats in the forced swim test at 5 nmol infused into the infralimbic cortex; induces anhedonia-like behavior in rats in a sucrose intake test at 6.25 nmol infused into the PFC; impairs memory acquisition, consolidation, and retrieval in mice in the novel object recognition test