Learn More
Abcam Dihydrokainic acid, EAAT2 (GLT-1) inhibitor, 10MG
Catalog No. 502584711
Encompass_Preferred
MW 215.25 Da. Selective, non-transportable inhibitor of glutamate transporter EAAT2 (GLT-1) (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4711 Supplier Abcam Supplier No. AB12006610MG
MW 215.25 Da. Selective, non-transportable inhibitor of glutamate transporter EAAT2 (GLT-1) (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM).
The product is subject to the following: Abcam Restricted Use Statement