Learn More
Abcam Dihydrokainic acid, EAAT2 (GLT-1) inhibitor, 50MG
Catalog No. 502584709
Encompass_Preferred
MW 215.25 Da. Selective, non-transportable inhibitor of glutamate transporter EAAT2 (GLT-1) (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4709 Supplier Abcam Supplier No. AB12006650MG
MW 215.25 Da. Selective, non-transportable inhibitor of glutamate transporter EAAT2 (GLT-1) (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM).
The product is subject to the following: Abcam Restricted Use Statement