Learn More
Medchemexpress LLC Dihydrokavain | 587-63-3 | 232.28 | 1 MG
SDP
Catalog No. 5000416281
Encompass_Preferred
Dihydrokavain (7,8-Dihydrokawain) is a natural kavalactone compound that inhibits COX-1, COX-2, CYP2C9 (IC50 = 130.95 μM), CYP2C19 (IC50 = 10.05 μM) and CYP3A4 (IC50 = 78.59 μM). It also reduces TNFα secretion and shows analgesic and anxiolytic effects.
  • Natural kavalactone compound.
  • Inhibits COX-1 and COX-2.
  • Inhibits CYP2C9 (IC50 = 130.95 μM), CYP2C19 (IC50 = 10.05 μM), and CYP3A4 (IC50 = 78.59 μM).
  • Reduces TNFα secretion.
  • Shows analgesic and anxiolytic effects.
Catalog No. 50-004-16281 Supplier Medchemexpress LLC Supplier No. HYN09201MG
Dihydrokavain (7,8-Dihydrokawain) is a natural kavalactone compound that inhibits COX-1, COX-2, CYP2C9 (IC50 = 130.95 μM), CYP2C19 (IC50 = 10.05 μM) and CYP3A4 (IC50 = 78.59 μM). It also reduces TNFα secretion and shows analgesic and anxiolytic effects.
  • Natural kavalactone compound.
  • Inhibits COX-1 and COX-2.
  • Inhibits CYP2C9 (IC50 = 130.95 μM), CYP2C19 (IC50 = 10.05 μM), and CYP3A4 (IC50 = 78.59 μM).
  • Reduces TNFα secretion.
  • Shows analgesic and anxiolytic effects.