Learn More
Cayman Chemical Dihydrokavain, Purity: ≥98%, CAS: 587-63-3, 5 mg
SDP
Catalog No. 502947880
Encompass_Preferred
An analytical reference standard categorized as a kavalactone; has analgesic and anxiolytic properties in mice and G. gallus chicks, respectively; intended for research and forensic applications
Catalog No. 50-294-7880 Supplier Cayman Chemical Supplier No. 338225 MG
An analytical reference standard categorized as a kavalactone; has analgesic and anxiolytic properties in mice and G. gallus chicks, respectively; intended for research and forensic applications