Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrolanosterol | 79-62-9 | 99.6% | 10 MG
SDP

Catalog No. 5000232126
Encompass_Preferred

Dihydrolanosterol is a sterol substrate of cytochrome P450 51 (CYP51) and a cholesterol biosynthesis inhibitor supplied as a purified analytical standard for research applications in sterol metabolism and enzymology.

  • Acts as a substrate of cytochrome P450 51 (CYP51).
  • Inhibits cholesterol biosynthesis, useful for pathway studies.
  • Supplied as a high-purity analytical standard (reported purity 99.56%).
  • Molecular formula C30H52O; molecular weight 428.73 g/mol.
  • Available in small, research-scale quantities such as 10 MG vials.
  • Store at 4°C protected from light; in solvent store at -80°C for long-term stability.
  • Certificate of analysis and safety data sheet available for verification.

Catalog No. 50-002-32126 Supplier Medchemexpress LLC Supplier No. HY12241010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrolanosterol is a sterol substrate of cytochrome P450 51 (CYP51) and a cholesterol biosynthesis inhibitor supplied as a purified analytical standard for research applications in sterol metabolism and enzymology.

  • Acts as a substrate of cytochrome P450 51 (CYP51).
  • Inhibits cholesterol biosynthesis, useful for pathway studies.
  • Supplied as a high-purity analytical standard (reported purity 99.56%).
  • Molecular formula C30H52O; molecular weight 428.73 g/mol.
  • Available in small, research-scale quantities such as 10 MG vials.
  • Store at 4°C protected from light; in solvent store at -80°C for long-term stability.
  • Certificate of analysis and safety data sheet available for verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.