Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrolanosterol | 79-62-9 | 99.6% | 5 MG
SDP

Catalog No. 5000233886
Encompass_Preferred

Dihydrolanosterol is a sterol research reagent that functions as a substrate of cytochrome P450 51 (CYP51) and inhibits cholesterol biosynthesis.

  • Acts as a CYP51 substrate and cholesterol biosynthesis inhibitor.
  • Reported purity 99.6%.
  • Molecular formula C30H52O; molecular weight 428.7 g/mol.
  • CAS number 79-62-9.
  • Supplied in small research quantities suitable for biochemical and enzymology studies.

Catalog No. 50-002-33886 Supplier Medchemexpress LLC Supplier No. HY1224105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrolanosterol is a sterol research reagent that functions as a substrate of cytochrome P450 51 (CYP51) and inhibits cholesterol biosynthesis.

  • Acts as a CYP51 substrate and cholesterol biosynthesis inhibitor.
  • Reported purity 99.6%.
  • Molecular formula C30H52O; molecular weight 428.7 g/mol.
  • CAS number 79-62-9.
  • Supplied in small research quantities suitable for biochemical and enzymology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.