Learn More
Medchemexpress LLC Dihydrolanosterol-d7 | 2260669-12-1 | 99.0% | 1 MG
SDP
Catalog No. 5000231288
Encompass_Preferred
Dihydrolanosterol-d7 is a deuterium-labeled analogue of dihydrolanosterol used as an isotope-labeled standard and tracer for mass spectrometry and metabolic studies. Dihydrolanosterol is a substrate of CYP51 and a cholesterol biosynthesis inhibitor.
  • Deuterium-labeled internal standard for LC-MS/MS and metabolic tracing.
  • Supports studies of cholesterol biosynthesis and CYP51 activity.
  • High reported purity suitable for analytical applications.
  • Supplied in small pack sizes for use as a reference standard.
Catalog No. 50-002-31288 Supplier Medchemexpress LLC Supplier No. HY122410S1MG
Dihydrolanosterol-d7 is a deuterium-labeled analogue of dihydrolanosterol used as an isotope-labeled standard and tracer for mass spectrometry and metabolic studies. Dihydrolanosterol is a substrate of CYP51 and a cholesterol biosynthesis inhibitor.
  • Deuterium-labeled internal standard for LC-MS/MS and metabolic tracing.
  • Supports studies of cholesterol biosynthesis and CYP51 activity.
  • High reported purity suitable for analytical applications.
  • Supplied in small pack sizes for use as a reference standard.