Learn More
Medchemexpress LLC Dihydrolinalool | 18479-51-1 | 98.1% | 156.27 | 100 G
SDP
Catalog No. 5000366385
Encompass_Preferred
Dihydrolinalool is commonly used as a flavor ingredient in various products, including food, beverages, and perfumes. It also serves as a base material for the synthesis of organic compounds like pharmaceuticals and agrochemicals. It has been studied for its potential use as a bio-based solvent due to its low toxicity and biodegradability, and for its antioxidant and anti-inflammatory properties.
  • Used as a flavor ingredient in food, beverages, and perfumes.
  • Serves as a base material for synthesizing pharmaceuticals and agrochemicals.
  • Studied as a bio-based solvent due to low toxicity and biodegradability.
  • Investigated for potential antioxidant and anti-inflammatory properties.
  • Functions as a biochemical reagent for life science research.
Catalog No. 50-003-66385 Supplier Medchemexpress LLC Supplier No. HYW010202100G
Dihydrolinalool is commonly used as a flavor ingredient in various products, including food, beverages, and perfumes. It also serves as a base material for the synthesis of organic compounds like pharmaceuticals and agrochemicals. It has been studied for its potential use as a bio-based solvent due to its low toxicity and biodegradability, and for its antioxidant and anti-inflammatory properties.
  • Used as a flavor ingredient in food, beverages, and perfumes.
  • Serves as a base material for synthesizing pharmaceuticals and agrochemicals.
  • Studied as a bio-based solvent due to low toxicity and biodegradability.
  • Investigated for potential antioxidant and anti-inflammatory properties.
  • Functions as a biochemical reagent for life science research.