Learn More
Medchemexpress LLC Dihydrolipoamide S-succinyltransferase (DLST) | >90.0% | 100 UG
SDP
Catalog No. 5000294359
Encompass_Preferred
Recombinant human dihydrolipoamide S-succinyltransferase (DLST) expressed in Sf9 insect cells and supplied as an N-terminal His-tagged, purified protein for research use. The product corresponds to residues D68-L453 (accession P36957), has an approximate molecular weight of 43.9 kDa, and is supplied as a 0.22 μm filtered solution in 20 mM Tris, 300 mM NaCl, pH 7.5, 10% glycerol. Purity is >90% (reducing SDS-PAGE), endotoxin is <1 EU/μg, shipped on dry ice, and stored at -80°C.
Catalog No. 50-002-94359 Supplier Medchemexpress LLC Supplier No. HYP75709100UG
Recombinant human dihydrolipoamide S-succinyltransferase (DLST) expressed in Sf9 insect cells and supplied as an N-terminal His-tagged, purified protein for research use. The product corresponds to residues D68-L453 (accession P36957), has an approximate molecular weight of 43.9 kDa, and is supplied as a 0.22 μm filtered solution in 20 mM Tris, 300 mM NaCl, pH 7.5, 10% glycerol. Purity is >90% (reducing SDS-PAGE), endotoxin is <1 EU/μg, shipped on dry ice, and stored at -80°C.