Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrolipoic Acid 100mg | 462-20-4 | 208.34 | C8H16O2S2 | 100 MG
SDP

Catalog No. 5000226442
Encompass_Preferred

Dihydrolipoic Acid (DHLA) is the reduced form of lipoic acid and functions as a thiol antioxidant and radical scavenger. It is supplied for research and analytical applications in solid and solution formats and is commonly provided as a 100 mg pack.

  • Potent antioxidant and radical scavenger.
  • Available as 100 mg solid and as 10 mM, 1 mL solutions in DMSO.
  • High listed purity (99.95%), suitable for analytical use.
  • Molecular weight 208.34 g/mol; formula C8H16O2S2.
  • Intended for research use only.

Catalog No. 50-002-26442 Supplier Medchemexpress LLC Supplier No. HY116807100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrolipoic Acid (DHLA) is the reduced form of lipoic acid and functions as a thiol antioxidant and radical scavenger. It is supplied for research and analytical applications in solid and solution formats and is commonly provided as a 100 mg pack.

  • Potent antioxidant and radical scavenger.
  • Available as 100 mg solid and as 10 mM, 1 mL solutions in DMSO.
  • High listed purity (99.95%), suitable for analytical use.
  • Molecular weight 208.34 g/mol; formula C8H16O2S2.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.