Learn More
Cayman Chemical Dihydrolipoic Acid, Purity: ≥95%, CAS: 462-20-4, 100 mg
SDP
Catalog No. 502969898
Encompass_Preferred
The reduced form of α-lipoic acid that acts as a general antioxidant at concentrations ranging from 0.01-0.5 mM; at 50-100 µM, triggers apoptosis of mouse embryonic stem cells and human cancer cells, suppressing proliferation
Catalog No. 50-296-9898 Supplier Cayman Chemical Supplier No. 16372100 MG
The reduced form of α-lipoic acid that acts as a general antioxidant at concentrations ranging from 0.01-0.5 mM; at 50-100 µM, triggers apoptosis of mouse embryonic stem cells and human cancer cells, suppressing proliferation