Learn More
Cayman Chemical DIhydrolIpoIc AcId 500mg
SDP
Catalog No. 502972320
Encompass_Preferred
The reduced form of α-lipoic acid that acts as a general antioxidant at concentrations ranging from 0.01-0.5 mM; at 50-100 µM, triggers apoptosis of mouse embryonic stem cells and human cancer cells, suppressing proliferation
Catalog No. 50-297-2320 Supplier Cayman Chemical Supplier No. 16372500 MG
Add to Cart
Add to Cart
The reduced form of α-lipoic acid that acts as a general antioxidant at concentrations ranging from 0.01-0.5 mM; at 50-100 µM, triggers apoptosis of mouse embryonic stem cells and human cancer cells, suppressing proliferation