Learn More
Medchemexpress LLC Dihydrolipoic Acid 50mg | 462-20-4 | 50 MG
SDP
Catalog No. 5000226869
Encompass_Preferred
Dihydrolipoic acid (DHLA) is the reduced form of α-lipoic acid and functions as a potent antioxidant. Supplied as a 50 mg research reagent, the product includes analytical documentation and safety information for handling and storage. Consult the provided certificates and safety data for experimental use.
  • Potent antioxidant and radical scavenger.
  • Provided as a 50 mg reagent suitable for research applications.
  • High purity (99.95%) with certificate of analysis and analytical data.
  • Includes SDS, H NMR, LC-MS, and RP-HPLC documentation.
  • Store solid at -20°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-26869 Supplier Medchemexpress LLC Supplier No. HY11680750MG
Dihydrolipoic acid (DHLA) is the reduced form of α-lipoic acid and functions as a potent antioxidant. Supplied as a 50 mg research reagent, the product includes analytical documentation and safety information for handling and storage. Consult the provided certificates and safety data for experimental use.
  • Potent antioxidant and radical scavenger.
  • Provided as a 50 mg reagent suitable for research applications.
  • High purity (99.95%) with certificate of analysis and analytical data.
  • Includes SDS, H NMR, LC-MS, and RP-HPLC documentation.
  • Store solid at -20°C; in solvent, store at -80°C for long-term stability.