Learn More
Cayman Chemical Dihydrolycorine, Purity: ≥98%, CAS: 6271-21-2, 10 mg
SDP
Catalog No. 502939005
Encompass_Preferred
A derivative of lycorine with antihypertensive and neuroprotective activities; inhibits methoxamine-induced contraction of isolated rabbit aortic rings and rat anococcygeus muscle (pA2s = 5.93 and 6.35, respectively); reduces MAP in anesthetized and conscious normotensive rats at 80 mg/kg; reduces electrically or phenylephrine-induced hypertension in pithed rats; reduces infarct size, cerebral edema, and myeloperoxidase activity in a rat model of focal cerebral ischemia-reperfusion injury when administered following reperfusion
Catalog No. 50-293-9005 Supplier Cayman Chemical Supplier No. 2780910 MG
A derivative of lycorine with antihypertensive and neuroprotective activities; inhibits methoxamine-induced contraction of isolated rabbit aortic rings and rat anococcygeus muscle (pA2s = 5.93 and 6.35, respectively); reduces MAP in anesthetized and conscious normotensive rats at 80 mg/kg; reduces electrically or phenylephrine-induced hypertension in pithed rats; reduces infarct size, cerebral edema, and myeloperoxidase activity in a rat model of focal cerebral ischemia-reperfusion injury when administered following reperfusion