Learn More
Cayman Chemical DIhydromethystIcIn 10mg
SDP
Catalog No. 502937862
Encompass_Preferred
A kavalactone; a P-gp inhibitor that increases calcein AM uptake by 50% in P388 mouse leukemia cancer cells overexpressing P-gp at 54.6 μM; has analgesic activity, increasing the latency to tail withdrawal in the tail-flick assay in mice at 275 mg/kg; decreases the infarct size in a mouse model of MCA-induced ischemia at 10 mg/kg
Catalog No. 50-293-7862 Supplier Cayman Chemical Supplier No. 2764410 MG
A kavalactone; a P-gp inhibitor that increases calcein AM uptake by 50% in P388 mouse leukemia cancer cells overexpressing P-gp at 54.6 μM; has analgesic activity, increasing the latency to tail withdrawal in the tail-flick assay in mice at 275 mg/kg; decreases the infarct size in a mouse model of MCA-induced ischemia at 10 mg/kg