Learn More
Medchemexpress LLC Dihydromethysticin | 19902-91-1 | 276.29 | 1 MG
SDP
Catalog No. 5000416946
Encompass_Preferred
Dihydromethysticin is an orally active natural ingredient extracted from Piper methysticum. It functions as an anti-cancer agent by inhibiting carboxylesterase 1 and CYP2A5, while also upregulating NLRC3 and inducing apoptosis. This compound has demonstrated anticancer activity against both colorectal cancer and lung adenoma.
  • Inhibits carboxylesterase 1 (Ki = 68.2 μM)
  • Inhibits CYP2A5
  • Upregulates NLRC3 and induces apoptosis
  • Exhibits anticancer activity against colorectal cancer and lung adenoma
  • Suppresses proliferation, migration, and invasion of colorectal cancer cells
  • Promotes G0/G1 arrest in colorectal cancer cells
  • Completely inhibits lung adenoma formation
Catalog No. 50-004-16946 Supplier Medchemexpress LLC Supplier No. HYN09211MG
Dihydromethysticin is an orally active natural ingredient extracted from Piper methysticum. It functions as an anti-cancer agent by inhibiting carboxylesterase 1 and CYP2A5, while also upregulating NLRC3 and inducing apoptosis. This compound has demonstrated anticancer activity against both colorectal cancer and lung adenoma.
  • Inhibits carboxylesterase 1 (Ki = 68.2 μM)
  • Inhibits CYP2A5
  • Upregulates NLRC3 and induces apoptosis
  • Exhibits anticancer activity against colorectal cancer and lung adenoma
  • Suppresses proliferation, migration, and invasion of colorectal cancer cells
  • Promotes G0/G1 arrest in colorectal cancer cells
  • Completely inhibits lung adenoma formation