Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydromyrcenol, 99.1%, MP Biomedicals™

Catalog No. ICN15090625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15090625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15090625 Supplier MP Biomedicals, Inc Supplier No. 0215090625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18479-58-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00004474
InChI Key XSNQECSCDATQEL-UHFFFAOYSA-N
Synonym dihydromyrcenol, 2,6-dimethyl-7-octen-2-ol, 7-octen-2-ol, 2,6-dimethyl, 1,1,5-trimethyl-6-heptenol, 1-2,6-dimethyloct-7-en-2-ol, dsstox_cid_9317, myrcenol, 6,10-dihydro, dsstox_rid_78761
PubChem CID 29096
ChEBI CHEBI:87528
IUPAC Name 2,6-dimethyloct-7-en-2-ol
SMILES CC(CCCC(C)(C)O)C=C

Specifications

Color Colorless
Quantity 25 g
Formula Weight 156.27
Physical Form Liquid
Chemical Name or Material Dihydromyrcenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.