Learn More
Apexbio Technology LLC Dihydromyricetin 27200-12-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063295
Encompass_Preferred
Dihydromyricetin also termed Ampelopsin is a natural flavanonol compound frequently investigated for its antioxidant and anticancer properties It exerts biological effects primarily through modulation of molecular pathways involving oxidative stress responses apoptosis regulation and GABA-A receptor enhancement at benzodiazepine binding sites Studies indicate dihydromyricetin acts as a positive allosteric modulator on GABA-A receptors suggesting potential for research related to alcohol intoxication and associated neural mechanisms In cellular assays dihydromyricetin demonstrates antiproliferative activity against various cancer cell lines typically exhibiting IC50 concentrations in the micromolar range Thus this compound is widely utilized in biomedical research including cell signaling pathway analysis antioxidant mechanism exploration and studies examining potential therapeutic roles in oncology and neuropharmacology
Catalog No. 50-000-63295 Supplier Apexbio Technology LLC Supplier No. N18495.1
Dihydromyricetin also termed Ampelopsin is a natural flavanonol compound frequently investigated for its antioxidant and anticancer properties It exerts biological effects primarily through modulation of molecular pathways involving oxidative stress responses apoptosis regulation and GABA-A receptor enhancement at benzodiazepine binding sites Studies indicate dihydromyricetin acts as a positive allosteric modulator on GABA-A receptors suggesting potential for research related to alcohol intoxication and associated neural mechanisms In cellular assays dihydromyricetin demonstrates antiproliferative activity against various cancer cell lines typically exhibiting IC50 concentrations in the micromolar range Thus this compound is widely utilized in biomedical research including cell signaling pathway analysis antioxidant mechanism exploration and studies examining potential therapeutic roles in oncology and neuropharmacology