Learn More
Apexbio Technology LLC Dihydromyricetin, 20mg. Cas: 27200-12-0 MFCD: MFCD00189451. Antioxidant and anti-cancer reagent
SDP
Supplier:  Apexbio Technology LLC N184920
Encompass_Preferred
Dihydromyricetin (Ampelopsin) is a flavanonol with antioxidant and anti-cancer activity,found to have anti-alcohol intoxication effects. Its anti-alcohol effects appear to be by its actions as a positive modulator of GABA-A receptors at the benzodiazepin Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5647