Learn More
Medchemexpress LLC HY-N0112 100mg , Dihydromyricetin Ampeloptin;Ampelopsin CAS:27200-12-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYN0112100MG
Encompass_Preferred
Medchemexpress, HY-N0112 100mg Dihydromyricetin Ampeloptin;Ampelopsin CAS:27200-12-0 Formula:C15H12O8 IC50: 48 μM (dihydropyrimidinase) mTORC1/2 Purity:98% Dihydromyricetin is a potent inhibitor with an IC 50 of 48 μM on dihydropyrimidinase . Dihydromyricetin can activate autophagy through inhibiting mTOR signaling. Dihydromyricetin suppresses the formation of mTOR complexes ( mTORC1 / 2 ). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3510